EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Chronic exposure of rat primary astrocyte cultures to manganese results in increased binding sites for the 'peripheral-type' benzodiazepine receptor ligand 3H-PK 11195.
PMID 10471200
Abstract
Alterations of 'peripheral-type' benzodiazepine receptors (PTBRs) in brain are a feature of hepatic encephalopathy (HE). Although ammonia toxicity has been implicated in the disorder, recent findings suggest an accumulation of manganese in the brains of cirrhotic patients dying in hepatic coma. In this study, we examined the expression of PTBRs as well as the binding of the selective PTBR ligand 3H-PK 11195 in cultured astrocytes following chronic exposure to manganese. When astrocytes were exposed to 100 microM manganese for 1 week, a 57% increase in Bmax for 3H-PK 11195 binding was detected (P < 0.01) with no change in the Kd value. However, an examination by RT-PCR of the expression of the isoquinoline-binding moiety of the PTBR complex in these cells revealed no change in PTBR mRNA levels following manganese treatment. These findings suggest that manganese up-regulates 3H-PK 11195 binding sites by a process which does not involve a change in transcription. In view of the proposed role of astrocytic PTBRs in 'neurosteroid' synthesis, manganese-induced increases of PTBRs could contribute to the pathogenesis of HE.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0424
PK 11195
C21H21ClN2O