EMAIL THIS PAGE TO A FRIEND
Journal of neurochemistry
Pituitary adenylate cyclase-activating polypeptide regulation of vasoactive intestinal polypeptide transcription requires Ca2+ influx and activation of the serine/threonine phosphatase calcineurin.
PMID 10501227
Abstract
A >15-fold increase in vasoactive intestinal polypeptide (VIP) mRNA and VIP peptide levels occurred in primary chromaffin cells following exposure to the neurotrophic neuropeptide pituitary adenylate cyclase-activating polypeptide (PACAP)-27 with an EC50 of approximately 2 nM. PACAP induction of VIP expression was blocked by methoxyverapamil or by a combination of nimodipine and omega-conotoxin MVIIC, indicating a requirement for PACAP-initiated calcium entry through voltage-dependent calcium channels for regulation of VIP biosynthesis. Ascomycin, which inhibits calcineurin through formation of an ascomycin/FKBP12/calcineurin ternary complex, abolished the PACAP-evoked increase in VIP expression, whereas rapamycin, which also binds to FKBP12 but does not cause inhibition of calcineurin, did not. Cyclosporin A, which inhibits calcineurin through formation of a cyclosporin A/cyclophilin/calcineurin complex, also abolished PACAP-evoked VIP biosynthesis. These data indicate that PACAP regulates the expression of VIP via a signaling pathway that requires calcium influx and activation of calcineurin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4188
ω-Conotoxin MVIIC, ≥95% (HPLC)
C106H178N40O32S7