EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Phenotypic distinction in Enterococcus faecium and Enterococcus faecalis strains between susceptibility and resistance to growth-enhancing antibiotics.
PMID 10508048
Abstract
Susceptibility of Enterococcus faecium and Enterococcus faecalis strains from animals and foods to growth-promoting antibiotics used in animal feed was tested by the agar dilution technique. Acquired resistance to bacitracin, narasin, tylosin, and virginiamycin was seen for both species, and for E. faecium, resistance to avilamycin and avoparcin was also seen. Drawing the distinction between susceptibility and resistance based on frequency distributions of MICs was easy with avoparcin, avilamycin, and tylosin but difficult with virginiamycin and to some extent also with bacitracin and narasin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1271
Narasin from Streptomyces auriofaciens, ≥98% (HPLC)
C43H72O11