EMAIL THIS PAGE TO A FRIEND
The American journal of physiology
Phosphorylation-dependent stimulation of prostanoid synthesis by nigericin in cerebral endothelial cells.
PMID 10516103
Abstract
Nigericin decreases intracellular pH (pH(i)) and stimulates prostanoid (PG) synthesis in endothelial cells from cerebral microvessels of newborn pigs. Nigericin-induced PG production was abolished by protein tyrosine kinase (PTK) inhibitors and amplified by phorbol 12-myristate 13-acetate (PMA) or protein tyrosine phosphatase (PTP) inhibitors. Nigericin-induced PG production in PMA-primed cells was potentiated by PTP inhibitors and abrogated by PTK inhibitors. Phospholipase A(2) (PLA(2)) activity was stimulated by nigericin in a phosphorylation-dependent manner. Nigericin's effects on PG production and PLA(2) activity were reproduced by ionomycin, which activates cytosolic PLA(2) (cPLA(2)). cPLA(2) was immunodetected in endothelial cell lysates. We found no evidence that nigericin's effects are mediated via mitogen-activated protein (MAP) kinase [extracellularly regulated kinase 1 (ERK1) and ERK2] activation: although nigericin stimulated detergent-soluble MAP kinase, its effects were not amplified by PMA or PTP inhibitors. Phosphorylation-dependent stimulation of PG synthesis was also observed when pH(i) was decreased by sodium propionate or a high level of CO(2). Altogether, our data indicate that nigericin and decreased pH(i) stimulate PG synthesis by a protein phosphorylation-dependent mechanism involving cross talk between pathways mediated by PTK and PTP and by protein kinase C; cPLA(2) appears to be a key enzyme affected by nigericin and decreased pH(i).
Related Materials