EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Somatostatin receptor interacting protein defines a novel family of multidomain proteins present in human and rodent brain.
PMID 10551867
Abstract
By using the yeast two-hybrid system we identified a novel protein from the human brain interacting with the C terminus of somatostatin receptor subtype 2. This protein termed somatostatin receptor interacting protein is characterized by a novel domain structure, consisting of six N-terminal ankyrin repeats followed by SH3 and PDZ domains, several proline-rich regions, and a C-terminal sterile alpha motif. It consists of 2185 amino acid residues encoded by a 9-kilobase pair mRNA; several splice variants have been detected in human and rat cDNA libraries. Sequence comparison suggests that the novel multidomain protein, together with cortactin-binding protein, forms a family of cytoskeletal anchoring proteins. Fractionation of rat brain membranes indicated that somatostatin receptor interacting protein is enriched in the postsynaptic density fraction. The interaction of somatostatin receptor subtype 2 with its interacting protein was verified by overlay assays and coimmunoprecipitation experiments from transfected human embryonic kidney cells. Somatostatin receptor subtype 2 and the interacting protein display a striking overlap of their expression patterns in the rat brain. Interestingly, in the hippocampus the mRNA for somatostatin receptor interacting protein was not confined to the cell bodies but was also observed in the molecular layer, suggesting a dendritic localization of this mRNA.