EMAIL THIS PAGE TO A FRIEND
Journal of neurochemistry
Dopamine neuronal transport kinetics and effects of amphetamine.
PMID 10582600
Abstract
The dopamine (DA) transporter (DAT) regulates DA neurotransmission by recycling DA back into neurons. Drugs that interfere with DAT function, e.g., cocaine and amphetamine, can have profound behavioral effects. The kinetics of DA transport by DAT in isolated synaptosomal or single cell preparations have been previously studied. To investigate how DA transport is regulated in intact tissue and to examine how amphetamine affects the DAT, the kinetics of DA uptake by the DAT were examined in tissue slices of the mouse caudate-putamen with fast-scan cyclic voltammetry. The data demonstrate that inward DA transport is saturable and sodium-dependent. Elevated levels of cytoplasmic DA resulting from disruption of vesicular storage by incubation with 10 microM Ro 4-1284 did not generate DA efflux or decrease its uptake rate. However, incubation with 10 microM amphetamine reduced the net DA uptake rate and increased extracellular DA levels due to DA efflux through the DAT. In addition, a new, elevated steady-state level of extracellular DA was established after electrically stimulated DA release in the presence of amphetamine, norepinephrine, and exogenous DA. These results from intact tissue are consistent with a kinetic model of the DAT established in more purified preparations in which amphetamine and other transported substances make the inwardly facing DAT available for outward transport of intracellular DA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H8502
Dopamine hydrochloride
C8H11NO2 · HCl