EMAIL THIS PAGE TO A FRIEND
Journal of biochemical and biophysical methods
Transfer of SDS-proteins from gel electrophoretic zones into mass spectrometry, using electroelution of the band into buffer without sectioning of the gel.
PMID 10647815
Abstract
Five SDS-proteins, ranging in molecular weight from 14 to 66 kDa, were detected without covalent fluorescent labeling by the automated gel electrophoresis apparatus with intermittent fluorescence scanning (HPGE apparatus, LabIntelligence) during electrophoresis in barbiturate buffer in the presence of Cascade Blue. The SDS-proteins were electroeluted from the gel into 220 microl of buffer by a modification of the procedure of Gombocz and Cortez. The electroeluate was freed of SDS, ultrafiltered and subjected to MALDI-TOF mass spectrometry. The masses of the five native proteins were found to be maintained after electrophoresis and electroelution in the presence of the potential contaminants SDS, barbituric acid and Cascade Blue. The procedure of protein transfer from SDS-PAGE into mass spectrometry, without excision of bands, gel maceration and protein recovery by diffusion, therefore is shown to be suitable for the identification by mass of intact proteins derived from gel electrophoretic bands.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L6385 α-Lactalbumin from bovine milk, For use as a marker in SDS-PAGE