EMAIL THIS PAGE TO A FRIEND
The Journal of general virology
Virus-encoded proteinases and proteolytic processing in the Nidovirales.
PMID 10725411
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11418467001 Chymotrypsin Sequencing Grade, from bovine pancreas