EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Preparation and pharmacological evaluation of novel glycoprotein (Gp) IIb/IIIa antagonists. 1. The selection of naphthalene derivatives.
PMID 10748713
Abstract
The synthesis and design using molecular modeling techniques for non-peptide, low molecular weight novel fibrinogen receptor (glycoprotein IIb/IIIa: Gp IIb/IIIa) antagonists, is reported. We used a highly potent serine protease inhibitor, Nafamostat, having an amidinonaphthyl unit as the starting compound. The compounds 4-(6-amidino-2-naphthylaminocarbonyl)phenoxyacetic acid (5a) and 4-(6-amidino-2-naphthalenecarboxamido)phenoxyacetic acid (5b) inhibited adenosin-5'-diphospate (ADP)-induced aggregation of human platelet-rich plasma (PRP) with IC50 values of 0.05 and 0.07 microM, respectively, and had lost their ability to inhibit a variety of serine proteases, including thrombin, factor Xa, plasmin and trypsin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
530263
6-Cyano-2-naphthol, 97%
C11H7NO