EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Crystal structure of a conformation-selective casein kinase-1 inhibitor.
PMID 10749871
Abstract
Members of the casein kinase-1 family of protein kinases play an essential role in cell regulation and disease pathogenesis. Unlike most protein kinases, they appear to function as constitutively active enzymes. As a result, selective pharmacological inhibitors can play an important role in dissection of casein kinase-1-dependent processes. To address this need, new small molecule inhibitors of casein kinase-1 acting through ATP-competitive and ATP-noncompetitive mechanisms were isolated on the basis of in vitro screening. Here we report the crystal structure of 3-[(2,4,6-trimethoxyphenyl) methylidenyl]-indolin-2-one (IC261), an ATP-competitive inhibitor with differential activity among casein kinase-1 isoforms, in complex with the catalytic domain of fission yeast casein kinase-1 refined to a crystallographic R-factor of 22.4% at 2.8 A resolution. The structure reveals that IC261 stabilizes casein kinase-1 in a conformation midway between nucleotide substrate liganded and nonliganded conformations. We propose that adoption of this conformation by casein kinase-1 family members stabilizes a delocalized network of side chain interactions and results in a decreased dissociation rate of inhibitor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O9808
2-Oxindole, 97%
C8H7NO
I0658
IC 261, ≥98% (HPLC)
C18H17NO4