EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Functional delineation of three groups of the ATP-dependent family of chromatin remodeling enzymes.
PMID 10779516
Abstract
ATP-dependent chromatin remodeling enzymes antagonize the inhibitory effects of chromatin. We compare six different remodeling complexes: ySWI/SNF, yRSC, hSWI/SNF, xMi-2, dCHRAC, and dNURF. We find that each complex uses similar amounts of ATP to remodel nucleosomal arrays at nearly identical rates. We also perform assays with arrays reconstituted with hyperacetylated or trypsinized histones and isolated histone (H3/H4)(2) tetramers. The results define three groups of the ATP-dependent family of remodeling enzymes. In addition we investigate the ability of an acidic activator to recruit remodeling complexes to nucleosomal arrays. We propose that ATP-dependent chromatin remodeling enzymes share a common reaction mechanism and that a key distinction between complexes is in their mode of regulation or recruitment.