EMAIL THIS PAGE TO A FRIEND
Organic letters
Multicomponent linchpin couplings of silyl dithianes: synthesis of the Schreiber C(16-28) trisacetonide subtarget for mycoticins A and B.
PMID 10836057
Abstract
[formula: see text] An efficient synthesis of trisacetonide (+)-11, the Schreiber C(16-28) subtarget for mycoticins A and B, is described. The key synthetic transformation entails a one-flask, five-component linchpin coupling tactic.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
514241
2-(tert-Butyldimethylsilyl)-1,3-dithiane, 97%
C10H22S2Si