EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis of chiral hydroxyl phospholanes from D-mannitol and their use in asymmetric catalytic reactions.
PMID 10843636
Abstract
Chiral hydroxyl monophosphane 3 [(2S,3S,4S,5S)-3,4-dihydroxy-2, 5-dimethyl-1-phenylphospholane] and bisphospholanes 5a [1,2-bis[(2S, 3S,4S,5S)-3,4-dihydroxy-2,5-dimethylphospholanyl]benzene] and 5b [1, 2-bis[(2S,3S,4S,5S)-2,5-diethyl-3,4-dihydroxyphospholanyl]benzene] were synthesized from readily available D-mannitol in high yields. Strategies for protection and deprotection of OH-groups in the presence of phosphines have been explored. Rate acceleration in the Baylis-Hillman reaction was observed when a hydroxyl phosphine was used as the catalyst. Rhodium complexes with chiral bisphospholanes are highly enantioselective catalysts for the asymmetric hydrogenation of various kinds of functionalized olefins such as dehydroamino acid derivatives, itaconic acid derivatives, and enamides. An interesting feature of the hydroxyl phospholane system is that hydrogenation of some substrates can be carried out in water with >99% ee and 100% conversion (e.g., itaconic acid).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17311
D-Mannitol, tested according to Ph.Eur.
C6H14O6