EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Physiological control of smooth muscle-specific gene expression through regulated nuclear translocation of serum response factor.
PMID 10866994
Abstract
Prolonged serum deprivation induces a structurally and functionally contractile phenotype in about 1/6 of cultured airway myocytes, which exhibit morphological elongation and accumulate abundant contractile apparatus-associated proteins. We tested the hypothesis that transcriptional activation of genes encoding these proteins accounts for their accumulation during this phenotypic transition by measuring the transcriptional activities of the murine SM22 and human smooth muscle myosin heavy chain promoters during transient transfection in subconfluent, serum fed or 7 day serum-deprived cultured canine tracheal smooth muscle cells. Contrary to our expectation, SM22 and smooth muscle myosin heavy chain promoter activities (but not viral murine sarcoma virus-long terminal repeat promoter activity) were decreased in long term serum-deprived myocytes by at least 8-fold. Because serum response factor (SRF) is a required transcriptional activator of these and other smooth muscle-specific promoters, we evaluated the expression and function of SRF in subconfluent and long term serum-deprived cells. Whole cell SRF mRNA and protein were maintained at high levels in serum-deprived myocytes, but SRF transcription-promoting activity, nuclear SRF binding to consensus CArG sequences, and nuclear SRF protein were reduced. Furthermore, immunocytochemistry revealed extranuclear redistribution of SRF in serum-deprived myocytes; nuclear localization of SRF was restored after serum refeeding. These results uncover a novel mechanism for physiological control of smooth muscle-specific gene expression through extranuclear redistribution of SRF and consequent down-regulation of its transcription-promoting activity.