EMAIL THIS PAGE TO A FRIEND
Hybridoma
A panel of CAb antibodies recognize endogenous and ectopically expressed ING1 protein.
PMID 10868796
Abstract
Nine monoclonal antibodies (MAbs) against human and rodent ING1 protein have been generated using an IL6-secreting mouse myeloma line. These antibodies are all effective in recognizing ING1 protein in ELISAs, Western blot assays, and by indirect immunofluorescence. Combining different CAb monoclonal antibodies in a Western blot assay also allows detection of the very low levels of endogenous ING1 found in fibroblast cells in culture and the identification of at least two isoforms of ING1 in normal human diploid fibroblasts and established brain cancer cell lines.