EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
EWS.Fli-1 fusion protein interacts with hyperphosphorylated RNA polymerase II and interferes with serine-arginine protein-mediated RNA splicing.
PMID 10982800
Abstract
Ewing's sarcoma displays a characteristic chromosomal translocation that results in fusion of the N-terminal domain of the Ewing's sarcoma protein (EWS) to the C-terminal DNA-binding domain of the ETS family transcription factor Fli-1 (Friend leukemia integration-1). EWS possesses structural motifs suggesting a role in transactivation as well as RNA binding. We demonstrate that wild-type EWS protein functions as an adapter molecule coupling transcription to RNA splicing by binding to hyperphosphorylated RNA polymerase II through the N-terminal domain of EWS and recruiting serine-arginine (SR) splicing factors through the C-terminal domain of EWS. The oncogenic EWS.Fli-1 fusion protein retains the ability to bind to hyperphosphorylated RNA polymerase II but lacks the ability to recruit SR proteins because of replacement of the C-terminal domain of EWS by Fli-1. In an in vivo splicing assay, the EWS.Fli-1 fusion protein inhibits SR protein-mediated E1A pre-mRNA splicing in a dominant-negative manner. These results indicate that EWS.Fli-1 interferes with the normal function of EWS and implicate uncoupling of gene transcription from RNA splicing in the pathogenesis of Ewing's sarcoma.