EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
A new substrate for the Biginelli cyclocondensation: direct preparation of 5-unsubstituted 3,4-dihydropyrimidin-2(1H)-ones from a beta-keto carboxylic acid
PMID 11052136
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
518050
3,5-Dimethylbenzaldehyde, 97%
C9H10O