EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Enzyme immobilization by adsorption on hydrophobic derivatives of cellulose and other hydrophilic materials.
PMID 1106329
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3272 Phenoxyacetylcellulose