EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Neuroprotection by group I metabotropic glutamate receptor antagonists in forebrain ischemia of gerbil.
PMID 11065123
Abstract
Stimulation of group I metabotropic glutamate receptors (mGluR 1 and 5) activates G-protein coupled-phospholipase C (PLC) to release 1,2-diacylglycerol (DAG) and arachidonic acid (ArAc). To elucidate the role of group I mGluR, we tested the effects of (S)-alpha-methyl-4-carboxy-phenylglycine (MCPG, mGluR 1 and 5 antagonist), 1-aminoindan-1,5-dicarboxylic acid (AIDA, mGluR 1a specific antagonist) and 2-methyl-6-(phenylethynyl) pyridine (MPEP, mGluR 5 antagonist) on ArAc release and neuronal survival after transient forebrain ischemia in gerbils. Ischemia resulted in (a) significant release of ArAc at 1-day reperfusion and (b) significant neuronal death in the hippocampal CA1 subfield after 6-day reperfusion. MCPG and MPEP decreased ArAc release and also significantly increased neuronal survival. AIDA was less effective in decreasing ArAc release and had no effect on neuronal death. These results suggest that activation of mGluR 5 may be an important pathway in ArAc release and neuronal death after transient ischemia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M4796
(±)-α-Methyl-(4-carboxyphenyl)glycine
C10H11NO4