EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Analysis of carnitine and acylcarnitines in urine by capillary electrophoresis.
PMID 11105876
Abstract
A capillary electrophoresis method is described for the simultaneous analysis of carnitine and short-chain acylcarnitines in aqueous standard solutions and urine samples. Samples were worked up using silica gel extraction and derivatization with 4'-bromophenacyl trifluoromethanesulfonate. Separation was performed in less than 8 min using a binary buffer system containing phosphate/phosphoric acid and sodium dodecyl sulfate. 3-(2,2,2-Trimethylhydrazinium)propionate (mildronate) was used as an internal standard. The method was developed with aqueous standard solutions and then applied successfully to spiked and unspiked human urine samples. The limit of detection for both carnitine and acetylcamitine is 3 microM.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
41392
4-Bromophenacyl trifluoromethanesulfonate, for HPLC derivatization, ≥95% (H-NMR/C-NMR)
C9H6BrF3O4S