EMAIL THIS PAGE TO A FRIEND
Chemistry & biology
Combinatorial synthesis of novel and potent inhibitors of NADH:ubiquinone oxidoreductase.
PMID 11137820
Abstract
NADH:ubiquinone oxidoreductase (complex I) is the first of three large enzyme complexes located in the cell's inner mitochondrial membrane which form the electron transport chain that carries electrons from NADH to molecular oxygen during oxidative phosphorylation. There is significant interest in developing small molecule inhibitors of this enzyme for use as biological probes, insecticides and potential chemopreventive/chemotherapeutic agents. Herein we describe the application of novel natural product-like libraries to the discovery of a family of potent benzopyran-based inhibitors. Initially a combinatorial library of benzopyrans, modeled after natural products, was synthesized using a solid phase cycloloading strategy. Screening of this diversity oriented library for inhibitory potency against NADH:ubiquinone oxidoreductase activity in vitro using bovine heart electron transport particles provided several lead compounds which were further refined through a series of focused libraries. Using this combinatorial library approach, a family of potent 2,2-dimethylbenzopyran-based inhibitors was developed with IC(50) values in the range of 18-55 nM. Cell-based assays revealed that these inhibitors were rather non-cytotoxic in the MCF-7 cell line; however, they were quite cytostatic in a panel of cancer cell lines suggesting their potential as chemotherapeutic/chemopreventive candidates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
452467
3,4-Dimethoxybenzenesulfonyl chloride, 98%
C8H9ClO4S