EMAIL THIS PAGE TO A FRIEND
Organic letters
Total synthesis of (+/-)-cytisine.
PMID 11150199
Abstract
[reaction:see text] The nicotine partial agonist cytisine was prepared in five steps featuring an "in situ" Stille or Suzuki biaryl pyridine coupling. Differentiation of the pyridyl rings was accomplished via selective benzylation and then reduction of a pyridinium ring. The penultimate diazabicyclo[3.3.1]nonane intermediate was obtained with high diastereoselectivity. A similar sequence has been employed for the synthesis of novel derivative 9-methoxycytisine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
712264
Cytisine, ≥99.0% (HPLC), ≥99%
C11H14N2O