EMAIL THIS PAGE TO A FRIEND
Phytochemistry
In vitro inhibition of Sphaeropsis sapinea by natural stilbenes.
PMID 11219808
Abstract
The effects of pinosylvin, pinosylvin monomethyl ether, pinosylvin dimethyl ether, and resveratrol on the fungal shoot blight and canker pathogen of conifers Sphaeropsis sapinea were examined in vitro. Effects of compounds, isolates, and concentrations on both conidial germination and mycelial growth were significant (values of P < 0.001), indicating inhibitory activity of these compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42914
Pinosylvin monomethyl ether, ≥97.0% (HPLC)
C15H14O2