EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacy and pharmacology
Simultaneous determination of (-)- and (+)- propranolol by gas chromatography-mass spectrometry using a deuterium labeling technique.
PMID 11325
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4875
D-Penicillamine, 98-101%
C5H11NO2S