EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and structure-activity studies of side-chain derivatized arylhydantoins for investigation as androgen receptor radioligands.
PMID 11327585
Abstract
A series of arylhydantoin derivatives modeled after the antiandrogen RU 58841 was generated to identify potential candidates for development as androgen receptor (AR) radioligands. Side-chain modified derivatives of RU 58841, suitable for labeling with either carbon-11 or radiohalogens (fluorine-18, iodine-123), were synthesized and tested for their AR binding affinities. The N-(iodopropenyl) derivative 13 (Ki = 13 nM) is a potential candidate for development as a radioiodinated AR ligand.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
515604
4-Iodobenzyl bromide, 95%
C7H6BrI