EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
A new mouse myeloma cell line that has lost immunoglobulin expression but permits the construction of antibody-secreting hybrid cell lines.
PMID 113458
Abstract
We have isolated a subclone of the mouse myeloma cell line P3-X63-Ag8 that does not express immunoglobulin heavy or light chains. This clone X63-Ag8.653 can be used for efficient fusion with antibody-forming cells to obtain hybrid cell lines producing pure monoclonal antibodies. Screening of hybrid cell lines for specificity and immunoglobulin classes was done with a modified enzyme-linked immunosorbent assay.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
85011420
P3X63Ag8.653 Cell Line from mouse