EMAIL THIS PAGE TO A FRIEND
Biochemical medicine
Glutaric aciduria; a "new" disorder of amino acid metabolism.
PMID 1137568
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72715
Glutaryl-L-carnitine lithium salt, ≥98.0% (TLC)
C12H21NO6 · xLi+