EMAIL THIS PAGE TO A FRIEND
FEBS letters
Isolation of large molecular weight DNA from agarose gels for further digestion by restriction enzymes.
PMID 1140401
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
60399
Potassium iodide, BioUltra, ≥99.5% (AT)
IK