EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
A mechanism-based affinity-labeling agent for possible use in isolating N-acetylglucosaminidase.
PMID 11425557
Abstract
We have prepared several mechanism-based affinity-labeling agents for possible use in isolating N-acetylglucosaminidase, in which an N-acetylglucosamine is linked to an o-monofluoro- or difluoro-methyl phenoxy glycoside with or without a cleavable disulfide group in the tether to biotin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4531
Biotin disulfide N-hydroxysuccinimide ester, >98%, powder
C22H33N5O7S3