EMAIL THIS PAGE TO A FRIEND
Cytogenetics and cell genetics
Assignment of the human metastasis-associated gene 1 (MTA1) to human chromosome band 14q32.3 by fluorescence in situ hybridization.
PMID 11474200