EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Identification and characterization of a novel inositol hexakisphosphate kinase.
PMID 11502751
Abstract
The inositol pyrophosphate disphosphoinositol pentakisphosphate (PP-InsP(3)/InsP(7)) is formed in mammals by two recently cloned inositol hexakiphosphate kinases, InsP(6)K1 and InsP(6)K2 (Saiardi, A., Erdjument-Bromage, H., Snowman, A. M., Tempst, P., and Snyder, S. H. (1999) Curr. Biol. 9, 1323-1326). We now report the identification, cloning, and characterization of a third InsP(7) forming enzyme designated InsP(6)K3. InsP(6)K3 displays 50 and 45% sequence identity to InsP(6)K1 and InsP(6)K2, respectively, with a smaller mass (46 kDa) and a more basic character than the other two enzymes. InsP(6)K3 is most enriched in the brain where its localization resembles InsP(6)K1 and InsP(6)K2. Intracellular disposition discriminates the three enzymes with InsP(6)K2 being exclusively nuclear, InsP(6)K3 predominating in the cytoplasm, and InsP(6)K1 displaying comparable nuclear and cytosolic densities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10726 CompoZr® Knockout ZFN Kit, Human IP6K3 (NM_001142883)