EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Gene expression profiling in human esophageal cancers using cDNA microarray.
PMID 11520067
Abstract
Human esophageal cancer cell lines and human esophageal cancer tissues were profiled on cDNA microarrays. In esophageal cancer cell lines, KYAE and OE-33 (adenocarcinomas) were distinguished from KYSE series (squamous cell carcinomas). Although SK-GT-4 and TE7 were derived from adenocarcinomas, they had a comparatively similar expression profile to the KYSE series. A set of genes whose expression commonly either increased or decreased in cancer cell lines was identified. Genes that were characteristically expressed in KYAE and OE-33 were also identified. The gene expression profiles of cancer tissues (CTs) were remarkably different from those of the cancer cell lines (CCLs). Notable differences between CCLs and CTs were observed in matrix metalloproteinases, plasminogen activator, collagens, paxillin, and thrombospondin 2, etc., whose expression was not increased in CCLs but increased in CTs. Twenty-three genes were extracted to categorize patients according to their prognoses, and clustering analyses, using these genes, were performed successfully.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11012002
KYAE-1 cell line