EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
A Practical Method for Epoxidation of Terminal Olefins with 30 Hydrogen Peroxide under Halide-Free Conditions.
PMID 11667823
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
02382
Methyltrioctylammonium hydrogen sulfate, ≥95.0% (T)
C25H55NO4S