EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Total Synthesis of (-)-Reserpine Using the Chiron Approach.
PMID 11671435
Abstract
A highly stereocontrolled synthesis of ring D/E precursor to reserpine has been developed starting from (-)-quinic acid as a chiral template. The total synthesis of (-)-reserpine is described through the cyclization of an immonium lactam intermediate.