EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Fluorination with XeF(2).(1) 44. Effect of Geometry and Heteroatom on the Regioselectivity of Fluorine Introduction into an Aromatic Ring.
PMID 11672087
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
394505
Xenon difluoride, 99.99% trace metals basis
F2Xe