EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
A New, Efficient Method for the Synthesis of Bisindolylmaleimides.
PMID 11672217
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
525898
Methyl (1-methylindolyl)-3-glyoxylate, 97%
C12H11NO3