EMAIL THIS PAGE TO A FRIEND
Laboratory animal science
Diagnostic exercise.
PMID 117225
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8010
(±)-Sulpiride
C15H23N3O4S