EMAIL THIS PAGE TO A FRIEND
Chemical reviews
Metal-catalyzed living radical polymerization.
PMID 11740919
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
673293
Pentamethylcyclopentadienylbis(triphenylphosphine)ruthenium(II) chloride
C46H45ClP2Ru