EMAIL THIS PAGE TO A FRIEND
Peptides
Interactions among challenges of hydromineral balance, angiotensin-converting enzyme, and cystine aminopeptidase.
PMID 11786201
Abstract
Enzymatic cleavage of some peptides could be included among the mechanisms of water-electrolyte homeostasis. To test this hypothesis, the angiotensin-converting activity (ACE) of plasma and the L-cystine-di-beta-naphthylamidase activity (CAP) of plasma and of soluble and particulate fractions from different areas of the central nervous system (CNS) were investigated in rats submitted to treatments eliciting hydromineral imbalance. CAP in the CNS was unchanged by hydromineral challenges. The correlations observed between plasma osmolality and CAP, and plasma CAP and ACE suggested a contribution of these activities to the restoration of basal water-electrolyte and blood pressure conditions through the hydrolysis of vasopressin, oxytocin, angiotensin I and bradykinin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5761
Glycine β-naphthylamide hydrochloride
C12H12N2O · HCl