EMAIL THIS PAGE TO A FRIEND
Chemical reviews
Room-Temperature Ionic Liquids. Solvents for Synthesis and Catalysis.
PMID 11849019
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
520659
Palladium(II) chloride, ≥99.9%
Cl2Pd