EMAIL THIS PAGE TO A FRIEND
Journal of combinatorial chemistry
Resin-capture and release strategy toward combinatorial libraries of 2,6,9-substituted purines.
PMID 11886295
Abstract
A resin-capture and release strategy for making combinatorial 2,6,9-trisubstituted purine libraries is demonstrated by capturing N9-derivatized purines at the C6 position with a thio-modified polymer. The C2 fluoro group is subsequently substituted with primary and secondary amines followed by thioether oxidation and release by C6 substitution with amines and anilines. This approach complements a previously reported strategy where a 6-phenylsulfenylpurine scaffold was captured at the C2 position with a resin-bound amine.(3)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
535125
2-Bromohypoxanthine, 96%
C5H3BrN4O