EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
First and unexpected synthesis of macrocyclic cyclophane-based unusual alpha-amino acid derivatives by phosphazene base without high dilution conditions.
PMID 11909729
Abstract
First synthesis of a macrocylic cyclophane-based unusual alpha-amino acid derivative 11 by coupling of ethyl isocyanoacetate with 1,2-bis(4-bromomethylphenyl)ethane under phase-transfer catalysis (PTC) conditions. Phosphazene base such as 2-tert-butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine (BEMP) is useful to improve the yield of cyclophane derivative without high dilution conditions.
Related Materials