EMAIL THIS PAGE TO A FRIEND
Organic letters
Transition metal salts-catalyzed aza-Michael reactions of enones with carbamates.
PMID 11950352
Abstract
Several transition metal salts were found to catalyze aza-Michael reactions of enones with carbamates efficiently. The catalytic activity was strongly dependent on the nature of the metal salts. While conventional Lewis acids such as BF(3).OEt(2), AlCl(3), or TiCl(4) showed lower activity, group 7-11 transition metal salts in higher oxidation states such as ReCl(5), Fe(ClO(4))(3).9H(2)O, RuCl(3).nH(2)O, OsCl(3).3H(2)O, RhCl(3).nH(2)O, PtCl(4).5H(2)O, or AuCl(3).H(2)O exhibited higher catalytic activity. [reaction: see text]
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
208523
Ruthenium(III) chloride, Ru content 45-55%
Cl3Ru