EMAIL THIS PAGE TO A FRIEND
Nutrition and cancer
Differential inhibition of human cancer cell proliferation by citrus limonoids.
PMID 11962254
Abstract
Limonoids have been shown to inhibit the growth of estrogen receptor-negative and -positive human breast cancer cells in culture. The primary objective of this study was to test the antiproliferative activity of limonoids (obacunone 17 beta-D-glucopyranoside, nomilinic acid 17 beta-D-glucopyranoside, limonin, nomilin, and a limonoid glucoside mixture), found in high concentrations in mandarin (Citrus reticulata Blanco), against a series of human cancer cell lines. The human cancer cell lines included leukemia (HL-60), ovary (SKOV-3), cervix (HeLa), stomach (NCI-SNU-1), liver (Hep G2), and breast (MCF-7). The growth-inhibitory effects of the four limonoids and the limonoid glucoside mixture against MCF-7 cells were significant, and the antiproliferative activity of the different citrus limonoids was also dose and time dependent. No significant effects were observed on growth of the other cancer cell lines treated with the four individual limonoids at 100 micrograms/ml. At 100 micrograms/ml, the limonoid glucoside mixture demonstrated a partial inhibitory effect on SKOV-3 cancer cells. With use of flow cytometry, it was found that all the limonoid samples could induce apoptosis in MCF-7 cells at relatively high concentrations (100 micrograms/ml). Considering the high concentration needed to induce apoptosis, it is unlikely that this is the primary mechanism of action for the cytotoxic effects seen with limonoids in this study. Further work is needed in this area to establish the mechanism of action of citrus limonoids on human breast cancer cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
64081
Limonin, analytical standard
C26H30O8
L9647
Limonin, from citrus seeds, >90% (HPLC)
C26H30O8