EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Ethyl alpha-fluoro silyl enol ether: stereoselective synthesis and its aldol reaction with aldehydes and ketones.
PMID 12003529
Abstract
Ethyl alpha-fluoro silyl enol ether is stereoselectively synthesized in high yield from inexpensive chlorofluoroacetate and Mg (or Zn) in DMF (or HMPA). Lewis acid promoted aldol reaction of this enol ether with aldehydes and ketones gives alpha-fluoro-beta-hydroxy esters in good to excellent yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
539147
Ethyl chlorofluoroacetate, 98%
C4H6ClFO2