EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Samarium(II) iodide-mediated intramolecular conjugate additions of alpha,beta-unsaturated lactones.
PMID 12027704
Abstract
Samarium(II) iodide, in the presence of catalytic amounts of nickel(II) iodide, has been used to promote intramolecular conjugate additions of alkyl halides onto alpha,beta-unsaturated lactones. This process has been shown to be applicable to a number of alpha,beta-unsaturated lactones, including tetrasubstituted olefins, and has been demonstrated to be quite general for the formation of saturated bicyclic and tricyclic lactones. The method presented herein provides a mild, efficient process to form structurally complex lactones from simple precursors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
446378
tert-Butyl(4-chlorobutoxy)dimethylsilane, 98%
C10H23ClOSi