EMAIL THIS PAGE TO A FRIEND
Nature
Betel nut constituents as inhibitors of gamma-aminobutyric acid uptake.
PMID 1207742
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G007
Guvacine hydrochloride, ≥97% (NMR), solid
C6H9NO2 · HCl