EMAIL THIS PAGE TO A FRIEND
Cancer research
Alterations in gene expression and activity during squamous cell carcinoma development.
PMID 12097286
Abstract
This study focuses on characterizing the genetic and biological alterations associated with squamous cell carcinoma development. Normal human epidermal keratinocytes (HEKs), cells isolated from a preneoplastic lesion (IEC-1), and two neoplastic cell lines, SCC-25 and COLO-16, were grown as raft cultures, and their gene expression profiles were screened using cDNA arrays. Our data indicated that the expression levels of at least 37 genes were significantly (P
Related Materials