EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Polyurea-encapsulated palladium(II) acetate: a robust and recyclable catalyst for use in conventional and supercritical media.
PMID 12122701
Abstract
Palladium(II) acetate microencapsulated in polyurea (MC-[Pd]) is an economical and versatile heterogeneous catalyst for a range of phosphine-free cross-coupling reactions in both conventional solvents and supercritical carbon dioxide (scCO2); the catalyst can be recovered by a simple filtration and recycled up to four times.