EMAIL THIS PAGE TO A FRIEND
Organic letters
Stereodivergent synthesis of enantiopure cis- and trans-3-Ethyl-4-piperidineacetates.
PMID 12153235
Abstract
[reaction: see text] Starting from a common chiral bicyclic lactam 11, enantiopure trans- or cis-3-ethyl-4-piperidineacetate derivatives are obtained by conjugate addition of an enolate or a cuprate to the unsaturated lactams 12 or 18, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43798
Ethyl 1,3-dithiolane-2-carboxylate, ≥98.0%
C6H10O2S2